missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Feline IL-18/IL-1F4 Recombinant Protein

Brand:  R&D Systems™ 5989-RM-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465624

  • £298.33 / 25µg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Measured by its ability to induce IFN-gamma secretion by KG-1 human acute myelogenous leukemia cells in the presence of TNF-alpha . The ED 50 for this effect is 0.1-0.5 µg/mL in the presence of 20 ng/mL rhTNF-alpha .
Specifications

Specifications

Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT.
IL-18/IL-1F4
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
18.3 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Feline
Unlabeled
Videos