missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Feline IL-12/IL-23 p40 Recombinant Protein

Brand:  R&D Systems™ 2117-FL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460604

  • £324.73 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ile23-Ser329 (Glu167Gly), with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 493741
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
36 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Feline
Yes
493741
IL-12/IL-23 p40
25μg
Unlabeled
Videos