missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Feline IFN-gamma Recombinant Protein

Brand:  R&D Systems™ 764-FG-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486264

  • £312.00 / 50µg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

Source: E. coli-derived, Gln24-Lys167, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 493965
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
17.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Feline
Yes
493965
IFN-gamma
50μg
Unlabeled
Videos