missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Feline IFN-gamma Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Gene ID (Entrez) | 493965 |
---|---|
Molecular Weight (g/mol) | 17.1 kDa |
Name | IFN-gamma |
Quantity | 50μg |
Source | E. coli-derived feline IFN-gamma protein Gln24-Lys167,with an N-terminal Met |
Description
The Recombinant Feline IFN-gamma Protein is derived from E. coli. The Recombinant Feline IFN-gamma Protein has been validated for the following applications: Bioactivity.Specifications
493965 | |
IFN-gamma | |
E. coli-derived feline IFN-gamma protein Gln24-Lys167,with an N-terminal Met | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
17.1 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |