missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Feline CXCL8/IL-8 Recombinant Protein

Brand:  R&D Systems™ 2277-FL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460924

  • £298.33 / 25µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala23-Ala101
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 493836
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Feline
Yes
493836
CXCL8/IL-8
25μg
Unlabeled
Videos