missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Equine IFN-gamma Recombinant Protein

Brand:  R&D Systems™ 1586-HG-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459733

  • £304.93 / 25µg


Estimated Shipment
03-09-2018
Add to basket

Description

Description

Source: E. coli-derived, Ala25-Gln166
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80.
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80 with BSA as a carrier protein.
IFN-gamma
25μg
Unlabeled
16.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Equine
Yes
Videos