missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ E. coli Endonuclease III/Buffer

Purified from E. coli containing a recombinant plasmid harboring the E. coli nth gene

Brand:  R&D Systems™ 4045-05K-EB

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13444014

  • £773.08 / 5000 units

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Endonucleases are enzymes that cleave nucleic acids (RNA or DNA) within the nucleotide strand. Endonuclease III is a DNA glycosylase with an associated AP lyase activity. Endonuclease III releases bases damaged by UV light, ionizing radiation, osmium tetroxide or acid. It cleaves abasic sites by b-elimination, producing a single nucleotide gap in the DNA, and contains an iron-sulfur group which helps to maintain its three dimensional conformation.

  • One Unit of enzyme cleaves 1 pmole of an oligonucleotide probe containing an AP site within an oligonucleotide duplex in one hour at 37°C
  • Catalyzes the excision of the following forms of DNA damage: Cis-trans-thymine glycol, 5,6-dihydrothymine, 5,6-dihydroxydihydrothymine, alloxan, urea, uracil, 5-hydroxy-5-methylhydantoin, methyltartronylurea, 6-hydroxy-5,6-dihydropyrimidines, 5-hydroxycytosine, 5-hydroxyuracil, 5-hydroxy-6-hydrothymine, 5,6-dihydrouracil, glycol, 5-hydroxy-6-hydrouracil, and AP sites
  • Suitable for use in FLARE™

Specifications

Specifications

E. coli Endonuclease III/Buffer
5,000U
Videos