Learn More
R&D Systems™ Cultrex Collagen I Cell Invasion Assay
24-well and 96-well kits that assess chemotaxis, cell migration, and/or cell invasion through a Collagen I matrix.
POA
Specifications
Type | Cultrex Collagen I Cell Invasion Assay |
---|---|
Includes | 5X Collagen I Solution, 10X Coating Buffer, 25X Cell Wash Buffer, 10X Cell Dissociation Solution, Calcein-AM |
Includes: 96 Well Cell Invasion Chamber; 5X Collagen I Solution, 1mL; 10X Coating Buffer, 1mL; 25X Cell Wash Buffer, 2 x 1.5mL; 10X Cell Dissociation Solution, 2 x 1.5mL; Calcein-AM, 50ug
Description
Collagens comprise a large family of insoluble extracellular glycoproteins that are essential components of connective tissues such as tendons, ligaments, cartilage, bone and skin. The mature polypeptides are secreted as coiled, left-handed helices that s
Flexible, standardized, high-throughput format for quantitating the degree to which invasive cells penetrate a barrier consisting of extracellular matrix protein Collagen I in response to chemoattractants and/or inhibiting compounds
Features a simplified
Specifications
Cultrex Collagen I Cell Invasion Assay | |
5X Collagen I Solution, 10X Coating Buffer, 25X Cell Wash Buffer, 10X Cell Dissociation Solution, Calcein-AM |