Learn More
R&D Systems™ Cultrex™ Collagen I Cell Invasion Assay
Reagent kit for investigating chemotaxis, cell migration, and/or cell invasion
Brand:  R&D Systems™ 3457-096-K
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443044
  • £391.31 / 1 set
Estimated Shipment
01-07-2019
Includes: 96 Well Cell Invasion Chamber; 5X Collagen I Solution, 1mL; 10X Coating Buffer, 1mL; 25X Cell Wash Buffer, 2 x 1.5mL; 10X Cell Dissociation Solution, 2 x 1.5mL; Calcein-AM, 50ug
Description
Description
Collagens comprise a large family of insoluble extracellular glycoproteins that are essential components of connective tissues such as tendons, ligaments, cartilage, bone and skin. The mature polypeptides are secreted as coiled, left-handed helices that s
Flexible, standardized, high-throughput format for quantitating the degree to which invasive cells penetrate a barrier consisting of extracellular matrix protein Collagen I in response to chemoattractants and/or inhibiting compounds
Features a simplified
Specifications
Specifications
5X Collagen I Solution, 10X Coating Buffer, 25X Cell Wash Buffer, 10X Cell Dissociation Solution, Calcein-AM
Each
The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Cultrex Collagen I Cell Invasion Assay
Videos