missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex Basement Membrane Extract, PathClear

Applications: Bioactivity

Brand:  R&D Systems™ 3432-005-01

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463004

  • £210.21 / 5mL

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Functional Assay: Tube formation - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling Assay: BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
Specifications

Specifications

Source: Murine Engelbreth-Holm-Swarm (EHS) tumor.
≤8EU/mL
Murine Engelbreth-Holm-Swarm tumor
12 - 18mg/mL
Bioactivity
Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
Videos
Documents