missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex 3-D Culture Matrix RGF Basement Membrane Extract

Applications: Bioactivity

Brand:  R&D Systems™ 3445-005-01

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13443034

  • £292.53 / 5mL

Estimated Shipment date
from Supplier 25-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Functional assays: Tube formation assay - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. 3D Culture - Basement Membrane Extract promotes differentiation of a human epithelial cell line derived from mammary gland (MCF-10A) or human prostate (PC-3) into acinar structures. Gelling assay: BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
Specifications

Specifications

14 to 16mg/mL
Bioactivity
Murine Engelbreth-Holm-Swarm (EHS) tumor.
≤8EU/mL
14 - 16mg/mL
Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
Videos
Documents