Learn More
R&D Systems™ CultreCoat™ BME Cell Invasion Assays, 96 well
Reagent kit for evaluating cell invasion for low, intermediary or highly invasive cell models
Brand:  R&D Systems™ 3482-096-K
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13433064
  • £527.73 / 96 tests
Estimated Shipment date
from Supplier 02-04-2020
Add to basket
Includes: 96 Well BME Cell Invasion Chamber (Low, Medium or High); 25X Cell Wash Buffer, 2 x 1.5mL; 10X Cell Dissociation Solution, 2 x 1.5mL; Calcein-AM, 50ug
Description
Description
Cell invasion is fundamental to angiogenesis, embryonic development, immune responses and tumor cell metastasis.
Contains one 96-well plate that is coated with BME, for cell models that exhibit a nominal invasive capacity: The low amount of BME coating provides the highest sensitivity for minimally invasive cell models
The medium amount of BME coating provides the
Specifications
Specifications
Medium BME Cell Invasion Assay
25X Cell Wash Buffer, 2 x 1.5mL; 10X Cell Dissociation Solution, 2 x 1.5mL; Calcein-AM, 50μg
1 Kit
The unopened product requires storage at both -20 to -70° C and 2 - 8° C. Consult the product insert for specific storage temperatures. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.