Learn More
R&D Systems™ CultreCoat™ BME Cell Invasion Assays, 96 well
Reagent kit for evaluating cell invasion for low, intermediary or highly invasive cell models
Brand:  R&D Systems™ 3483-096-K
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443064
  • £457.62 / 96 tests
Estimated Shipment
28-06-2019
Includes: 96 Well BME Cell Invasion Chamber (Low, Medium or High); 25X Cell Wash Buffer, 2 x 1.5mL; 10X Cell Dissociation Solution, 2 x 1.5mL; Calcein-AM, 50ug
Description
Description
Cell invasion is fundamental to angiogenesis, embryonic development, immune responses and tumor cell metastasis.
Contains one 96-well plate that is coated with BME, for cell models that exhibit a nominal invasive capacity: The low amount of BME coating provides the highest sensitivity for minimally invasive cell models
The medium amount of BME coating provides the
Specifications
Specifications
25X Cell Wash Buffer, 2 x 1.5mL; 10X Cell Dissociation Solution, 2 x 1.5mL; Calcein-AM, 50μg
1 Kit
The unopened product requires storage at both -20 to -70° C and 2 - 8° C. Consult the product insert for specific storage temperatures. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
High BME Cell Invasion Assay
Videos