missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cotton Rat IFN-gamma Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 565-RG-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13434024

  • £323.40 / 25µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Cotton Rat IFN-gamma Protein is derived from E. coli. The Recombinant Cotton Rat IFN-gamma Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in PBS, NaCl and Sucrose.
IFN-gamma
25μg
3 months from date of receipt, 2 to 8° C as supplied.
Yes
17 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Cotton Rat
Unlabeled
Videos
Documents