missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cotton Rat IFN-gamma Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Molecular Weight (g/mol) | 17 kDa |
Name | IFN-gamma |
Quantity | 25μg |
Recombinant | Yes |
Description
The Recombinant Cotton Rat IFN-gamma Protein is derived from E. coli. The Recombinant Cotton Rat IFN-gamma Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
IFN-gamma | |
Yes |
17 kDa | |
25μg | |
E. coli-derived cotton rat IFN-gamma protein His21-Ile170,with an N-terminal Met and two substitutions: G150R,Y165N |