missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cotton Rat IFN-alpha (Cys109Tyr) Recombinant Protein

Brand:  R&D Systems™ 1082-CR-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458523

  • £290.41 / 25µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Cys24-Lys190 (Cys109Tyr)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
IFN-alpha (Cys109Tyr)
25μg
Unlabeled
19.4 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Cotton Rat
Yes
Videos