missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cotton Rat IFN-alpha (Cys109Tyr) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Molecular Weight (g/mol) | 19.4 kDa |
---|---|
Name | IFN-alpha (Cys109Tyr) |
Quantity | 25μg |
Source | E. coli-derived cotton rat IFN-alpha protein Cys24-Lys190 (Cys109Tyr) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Description
The Recombinant Cotton Rat IFN-alpha (Cys109Tyr) Protein is derived from E. coli. The Recombinant Cotton Rat IFN-alpha (Cys109Tyr) Protein has been validated for the following applications: Bioactivity.Specifications
19.4 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
IFN-alpha (Cys109Tyr) | |
E. coli-derived cotton rat IFN-alpha protein Cys24-Lys190 (Cys109Tyr) | |
IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D | |
Recombinant |