missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cotton Rat CXCL1/2/3/GRO Recombinant Protein

Brand:  R&D Systems™ 1114-GR-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488563

  • £347.17 / 25µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Source: E. coli-derived, Ala28- Lys100
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
CXCL1/2/3/GRO
25μg
Unlabeled
7.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Cotton Rat
Yes
Videos