missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cotton Rat CCL3/MIP-1 alpha Recombinant Protein

Brand:  R&D Systems™ 1061-RM-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488413

  • £352.00 / 25µg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

Source: E. coli-derived, Ala24-Ala92
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
CCL3/MIP-1 alpha
25μg
Unlabeled
7.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Cotton Rat
Yes
Videos