missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CometAssay™ Single Cell Gel Electrophoresis Assay

Provides a simple and effective method for evaluating DNA damage in cells

Brand:  R&D Systems™ 4250-050-K

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13464434

  • £332.22 / 50 tests

Estimated Shipment date
from Supplier 05-03-2020

Please to purchase this item. Need a web account? Register with us today!


Includes: Lysis Solution, 2 x 500mL; Comet LMAgarose, 15mL; Trevigen CometSlide, 25/kit; pH 10 200mM EDTA, 12.5mL; Kit with Silver Stain (Mfr. No. 4251-050-K) also includes four staining reagents and a fixation additive.

Description

Description

The principle of the assay is based upon the ability of denatured, cleaved DNA fragments to migrate out of the nucleoid under the influence of an electric field, whereas undamaged DNA migrates slower and remains within the confines of the nucleoid when a

Choose the Neutral CometAssay to detect double-stranded breaks or the Alkaline CometAssay for a more sensitive protocol used to detect smaller amounts of damage including single and double-stranded breaks
Cells are immobilized in a bed of low melting poi

Specifications

Specifications

50 Tests
Single Cell Gel Electrophoresis Assay
Lysis Solution (2 x 500mL), Comet LMAgarose (15mL); Trevigen CometSlide (25/kit) and pH 10 200mM EDTA (12.5mL)
The product requires storage at both 2 - 8° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Videos
Documents