missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Chicken Netrin-2 Recombinant Protein

Brand:  R&D Systems™ 127-N2-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13489043

  • £347.17 / 25µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ala16-Pro581, with a C-terminal 6-His tag & Ala16-Pro581, with an N-terminal Met and C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 64 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Entrez Gene ID: 396387
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
64 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Chicken
Yes
396387
Netrin-2
25μg
Unlabeled
Videos