missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Chicken Netrin-1 Recombinant Protein

Brand:  R&D Systems™ 128-N1-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459063

  • £324.73 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Gly26-Ala606, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 66.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 396389
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
66.3 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Chicken
Yes
396389
Netrin-1
25μg
Unlabeled
Videos