missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Canine VEGF Recombinant Protein

Brand:  R&D Systems™ 1603-CV-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459773

  • £364.33 / 10µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Pro28-Arg190
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 403802
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
19 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Canine
Unlabeled
403802
VEGF
10μg
Disulfide-linked homodimer
Yes
Videos