missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Canine TNF-alpha Recombinant Protein

Brand:  R&D Systems™ 1507-CT-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459573

  • £340.57 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Val77-Leu233, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 403922
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
17.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Canine
Yes
403922
TNF-alpha
25μg
Unlabeled
Videos