missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Canine IL-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 403785 |
---|---|
Molecular Weight (g/mol) | 12.9 kDa |
Name | IL-4 |
Quantity | 25μg |
Source | E. coli-derived canine IL-4 protein His25-His132,with an N-terminal Met |
Description
The Recombinant Canine IL-4 Protein is derived from E. coli. The Recombinant Canine IL-4 Protein has been validated for the following applications: Bioactivity.Specifications
403785 | |
IL-4 | |
E. coli-derived canine IL-4 protein His25-His132,with an N-terminal Met | |
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Recombinant |
12.9 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |