missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Canine IL-21 Recombinant Protein

Brand:  R&D Systems™ 5849-ML-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465574

  • £316.81 / 25µg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Measured in a cell proliferation assay using N1186 human T cells. Parrish-Novak, J. et al. (2000) Nature 408:57. The ED 50 for this effect is 1-5 ng/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
15.0 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Canine
Unlabeled
442935
IL-21
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos