missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Canine IFN-gamma Recombinant Protein

Brand:  R&D Systems™ 781-CG-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486304

  • £309.00 / 50µg


Estimated Shipment
28-06-2018
Add to basket

Description

Description

Source: E. coli-derived, Gln24-Lys166, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 403801
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
17 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Canine
Yes
403801
IFN-gamma
50μg
Unlabeled
Videos