missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Canine IFN-gamma ELISpot Kit

For the quantitative determination of the frequency of cells releasing canine IL-4.

Brand:  R&D Systems™ EL781

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465125

  • £390.73 / 1 set


Estimated Shipment
03-04-2019
Add to basket

Includes:
  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control

Description

Description

Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells
Specifications

Specifications

IFN-gamma
Detection of cytokine secreting cells, quantitative sandwich ELISA
Canine IFN-gamma ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Canine
Canine IFN-gamma Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Canine IFN-gamma Positive Control
Whole Cells
1 x 96-well plate
Videos