missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Canine HGF Recombinant Protein

Brand:  R&D Systems™ 3386-HG-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462924

  • £316.81 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Arg28-Arg494 (Lys266Gln, Trp321Arg, Leu458Pro) (a chain) & Val495-Ser730 (Ile693Thr) (β chain)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 403441
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
HGF
25μg
Disulfide-linked heterodimer
Yes
403441
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Canine
Unlabeled
Videos