missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Canine GM-CSF Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1546-GM-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449653

  • £410.13 / 25µg

Estimated Shipment date
from Supplier 28-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Canine GM-CSF Protein is derived from E. coli. The Recombinant Canine GM-CSF Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
14.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Canine
<0.01 EU per 1μg of the protein by the LAL method.
Unlabeled
403923
GM-CSF
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Yes
Videos
Documents