missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Canine GM-CSF Recombinant Protein

Brand:  R&D Systems™ 1546-GM-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459653

  • £368.29 / 25µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala18-Lys144
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 403923
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
14.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Canine
Unlabeled
403923
GM-CSF
25μg
<0.01 EU per 1μg of the protein by the LAL method.
Yes
Videos