missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Canine CXCL8/IL-8 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 403850 |
Molecular Weight (g/mol) | 9 kDa (mature), 8.5 kDa (N-terminal truncation) |
Name | CXCL8/IL-8 |
Quantity | 25μg |
Description
The Recombinant Canine IL-8/CXCL8 Protein is derived from E. coli. The Recombinant Canine IL-8/CXCL8 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
9 kDa (mature), 8.5 kDa (N-terminal truncation) | |
25μg | |
E. coli-derived canine IL-8/CXCL8 protein Ala23-Pro101 & Val28-Pro101 |
403850 | |
CXCL8/IL-8 | |
Yes |