missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Bovine Osteopontin/OPN Protein

Brand:  R&D Systems™ 109-OP-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488483

  • £261.00 / 50µg


Estimated Shipment
28-06-2018
Add to basket

Description

Description

Source: Bovine milk-derived
Purity: 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 281499
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Osteopontin/OPN
50μg
Unlabeled
281499
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Bovine
Videos