Learn More
R&D Systems™ Bovine IL-5 Recombinant Protein
Brand: R&D Systems™ 2525-BL-025
Code : Z1
Additional Details : Weight : 0.00010kg
Description
Source: Mouse myeloma cell line, NS0-derived, Asn20-Ser134, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 280825
Regulatory Status: RUO
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
14 kDa (monomer) | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. | |
Bovine | |
Unlabeled |
280825 | |
IL-5 | |
25μg | |
Disulfide-linked homodimer | |
Yes |