missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Bovine IL-2 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 280822 |
Molecular Weight (g/mol) | 15.6 kDa |
Name | IL-2 |
Quantity | 20μg |
Description
The Recombinant Bovine IL-2 Protein is derived from E. coli. The Recombinant Bovine IL-2 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
15.6 kDa | |
20μg | |
E. coli-derived bovine IL-2 protein Ala21-Thr155,with an N-terminal Met |
280822 | |
IL-2 | |
Yes |