missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Bovine Fibronectin Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1030-FN-01M

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438343

  • £173.46 / 1mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Bovine Fibronectin Protein is derived from Bovine Plasma. The Bovine Fibronectin Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Tris-HCl, NaCl and Urea.
Fibronectin
1mg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 6 months, 2 to 8° C under sterile conditions after opening. Avoid vortexing and excessive agitation.
280794
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Bovine
Unlabeled
Videos
Documents