missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Bovine FGF acidic Protein

Brand:  R&D Systems™ 132-FA-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459163

  • £324.73 / 25µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: Bovine brain tissue-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Entrez Gene ID: 281160
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein.
FGF acidic
25μg
Unlabeled
281160
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Bovine
Videos