missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Amphibian TGF-beta 5 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 397778 |
Molecular Weight (g/mol) | 12.6 kDa |
Name | TGF-beta 5 |
Recombinant | Yes |
Description
The Recombinant Amphibian TGF-beta 5 Protein is derived from Sf 21 (baculovirus). The Recombinant Amphibian TGF-beta 5 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
12.6 kDa | |
Yes |
397778 | |
TGF-beta 5 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived amphibian TGF-beta 5 protein Gly271-Ser382 & Gln274-Ser382 |