missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Amphibian TGF-beta 5 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 245-B5-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13441254

  • £733.40 / 10µg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Amphibian TGF-beta 5 Protein is derived from Sf 21 (baculovirus). The Recombinant Amphibian TGF-beta 5 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
12.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Amphibian
Unlabeled
Reconstitute at 20 μg/mL in sterile 4 mM HCl containing 0.1% human or bovine serum albumin,
397778
TGF-beta 5
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos