missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%(e.e.>99.5%), Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
Specifications
Melting Point | 46.0°C to 49.0°C |
Color | White |
Boiling Point | 116.0°C to 118.0°C (1.5 mmHg) |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | C6H5C(OCH3)(CF3)CO2H |
Quantity | 1 g |
Fieser | 11,329 |
Merck Index | 15,6365 |
Show More |
Safety and Handling
Product Identifier
- (R)-(+)-a-Methoxy-a-(trifluoromethyl)phenylacetic acid
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'