missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%(e.e.>99.5%), Thermo Scientific Chemicals

Catalog Number p-2689255
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20445-31-2
C10H9F3O3
234.17
MFCD00004184
JJYKJUXBWFATTE-UHFFFAOYNA-N
+-mtpa, unii-27o5l9t1wm, r-moshers acid, +-mosher's acid, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-alpha-methoxy-alpha-trifluoromethylphenylacetic acid, r-mtpa, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-a-methoxy-a-trifluoromethylphenylacetic acid, r-3,3,3-trifluoro-2-methoxy-2-phenyl-propionic acid
2723917
(2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
COC(C(O)=O)(C1=CC=CC=C1)C(F)(F)F
This item is not returnable. View return policy
20445-31-2
C10H9F3O3
234.17
MFCD00004184
JJYKJUXBWFATTE-UHFFFAOYNA-N
+-mtpa, unii-27o5l9t1wm, r-moshers acid, +-mosher's acid, 2r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-alpha-methoxy-alpha-trifluoromethylphenylacetic acid, r-mtpa, r-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, r-+-a-methoxy-a-trifluoromethylphenylacetic acid, r-3,3,3-trifluoro-2-methoxy-2-phenyl-propionic acid
2723917
(2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
COC(C(O)=O)(C1=CC=CC=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY
Melting Point 46.0°C to 49.0°C
Color White
Boiling Point 116.0°C to 118.0°C (1.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5C(OCH3)(CF3)CO2H
Quantity 250 mg
Fieser 11,329
Merck Index 15,6365
Specific Rotation + 69.00
Formula Weight 234.17
Percent Purity 99%(e.e.>99.5%)
Physical Form Crystalline Low Melting Solid
Chemical Name or Material (R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, 99%(e.e.>99.5%)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'