missing translation for 'onlineSavingsMsg'
Learn More

(R)-α-Allylalanine hydrate, 98%, 98% ee, Thermo Scientific™

Catalog Number p-4258739
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
1266371-17-8, 96886-56-5
C6H11NO2·xH2O
129.16
(R)-(+)-2-Amino-2-methyl-4-pentenoic acid hydrate
This item is not returnable. View return policy
1266371-17-8, 96886-56-5
C6H11NO2·xH2O
129.16
(R)-(+)-2-Amino-2-methyl-4-pentenoic acid hydrate
CAS Min % 97.5
CAS Max % 100.0
Melting Point 271°C to 276°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (as Monohydrate)
Quantity 100 mg
Specific Rotation 26
Formula Weight 129.16
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-α-Allylalanine hydrate, 98% ee
Show More Show Less
Product Identifier
  • (R)-alpha-Allylalanine hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'