missing translation for 'onlineSavingsMsg'
Learn More

R-96544 hydrochloride, Tocris Bioscience™

Potent, selective 5-HT2A antagonist

Brand:  Tocris (Bio-Techne) 174250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13429733

  • £689.43 / 50mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective 5-HT2 receptor antagonist; displays some selectivity for 5-HT2A receptors (Ki = 1.6 nM). IC50 values are 2.2, 310, 2400, 3700, > 5000 and > 5000 nM for 5-HT2, α1-adrenergic, D2 dopamine, 5-HT1, 5-HT3 and β-adrenergic receptors respectively. Inhibits 5-HT-induced platelet aggregation and pressor responses in vivo.
Specifications

Chemical Identifiers

167144-80-1
391.936
2r,4r-5-2-2-2-3-methoxyphenyl ethyl phenoxy ethyl-1-methyl-3-pyrrolidinol hydrochloride, 3-pyrrolidinol,5-2-2-2-3-methoxyphenyl ethyl phenoxy ethyl-1-methyl-, 3r,5r, 2r,4r-4-hydroxy-2-2-2-2-3-methoxyphenyl ethyl phenoxy ethyl-1-methylpyrrolidine hydrochloride, 3r,5r-5-2-2-2-3-methoxyphenyl ethyl phenoxy ethyl-1-methylpyrrolidin-3-ol hydrochloride
(3R,5R)-5-[2-[2-[2-(3-methoxyphenyl)ethyl]phenoxy]ethyl]-1-methylpyrrolidin-3-ol;hydrochloride
C22H30ClNO3
OUKRSTJUAQEATQ-GZJHNZOKSA-N
16759150
CN1CC(CC1CCOC2=CC=CC=C2CCC3=CC(=CC=C3)OC)O.Cl

Specifications

50mg
167144-80-1
2r,4r-5-2-2-2-3-methoxyphenyl ethyl phenoxy ethyl-1-methyl-3-pyrrolidinol hydrochloride, 3-pyrrolidinol,5-2-2-2-3-methoxyphenyl ethyl phenoxy ethyl-1-methyl-, 3r,5r, 2r,4r-4-hydroxy-2-2-2-2-3-methoxyphenyl ethyl phenoxy ethyl-1-methylpyrrolidine hydrochloride, 3r,5r-5-2-2-2-3-methoxyphenyl ethyl phenoxy ethyl-1-methylpyrrolidin-3-ol hydrochloride
CN1CC(CC1CCOC2=CC=CC=C2CCC3=CC(=CC=C3)OC)O.Cl
391.936
391.94
R-96544 hydrochloride
C22H30ClNO3
OUKRSTJUAQEATQ-GZJHNZOKSA-N
(3R,5R)-5-[2-[2-[2-(3-methoxyphenyl)ethyl]phenoxy]ethyl]-1-methylpyrrolidin-3-ol;hydrochloride
16759150
>99%
Videos
Documents