Learn More
Tocris Bioscience™ R 59-022
Diacylglycerol kinase inhibitor; increases PKC activity
Brand:  Tocris Bioscience™ 219410
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13460764
  • £133.65 / 10mg
Estimated Shipment
02-01-2019
Zum Warenkorb hinzuf├╝gen
Beschreibung
Beschreibung
Diacylglycerol (DAG) kinase inhibitor (IC50 = 2.8 μM); increases protein kinase C activity. Potentiates thrombin-induced platelet aggregation and induces neutrophil chemotaxis. Inhibits U46619-induced contractions in mouse aorta and porcine coronary artery.
Spezifikation
Spezifikation
R 59-022
>98
10mg
R 59-022
459.58