missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-(-)-4-Fluorostyrene oxide, ChiPros 98+%, ee 99%

Brand:  Alfa Aesar™ L19583.06

Code : NEW

Additional Details : CAS Number : 134356-73-3 Weight : 0.00500kg Transport : UN number : 1992 Chem class : 3 Pack group : III

Quantity 5g

Product Code. 11355355

  • £355.10 / 5g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Specifications

(R)-(-)-4-Fluorostyrene oxide
99%
C8H7FO
MFCD03788740
4350328
92°C (14mmHg)
77°C (171°F)
134356-73-3
ee 99%
138.14
UN1992
(R)-2-(4-Fluorophenyl)oxirane
5g
−17° (c=1 in Chloroform)
Videos
SDS