missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-4-fluoro-α-methylbenzyl alcohol, ACROS Organics™

Brand:  Acros Organics 375450010

Code : 29

Additional Details : CAS Number : 101219-68-5 Weight : 0.05100kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 10007012

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

101219-68-5
140.157
PSDSORRYQPTKSV-ZCFIWIBFSA-N
852998
CC(C1=CC=C(C=C1)F)O
C8H9FO
MFCD03092997
r-1-4-fluorophenyl ethanol, benzenemethanol,4-fluoro-a-methyl-, ar, 1r-1-4-fluorophenyl ethanol, 1r-1-4-fluorophenyl ethan-1-ol, r-4-fluoro-alpha-methylbenzyl alcohol, pubchem14245, r-4-fluorophenylethan-1-ol, r-p-fluoro-alpha-methylbenzylalcohol, r-+-1-4-fluorophenyl ethanol, r-4-fluoro-, a-methylbenzyl alcohol
(1R)-1-(4-fluorophenyl)ethanol

Specifications

(R)-(+)-4-Fluoro-α-methylbenzyl alcohol
Authentic
Colorless to Yellow
101219-68-5
CH3CH(OH)C6H4F
r-1-4-fluorophenyl ethanol, benzenemethanol,4-fluoro-a-methyl-, ar, 1r-1-4-fluorophenyl ethanol, 1r-1-4-fluorophenyl ethan-1-ol, r-4-fluoro-alpha-methylbenzyl alcohol, pubchem14245, r-4-fluorophenylethan-1-ol, r-p-fluoro-alpha-methylbenzylalcohol, r-+-1-4-fluorophenyl ethanol, r-4-fluoro-, a-methylbenzyl alcohol
CC(C1=CC=C(C=C1)F)O
140.157
140.16
≥94%
97.5% min. (GC)
Glass bottle
1g
C8H9FO
MFCD03092997
PSDSORRYQPTKSV-ZCFIWIBFSA-N
(1R)-1-(4-fluorophenyl)ethanol
852998
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (R)-(+)-4-Fluoro-a-methylbenzyl alcohol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS