missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane, 98%, Thermo Scientific™

Catalog Number p-3770993
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
57044-24-3
C6H11ClO2
150.60
MFCD00273371
BNPOTXLWPZOESZ-UHFFFAOYNA-N
r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, r-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, 4r, r-3-chloro-1,2-propanediol acetonide, pubchem14074, ksc911s2r
2734443
(4R)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
CC1(C)OCC(CCl)O1
This item is not returnable. View return policy
57044-24-3
C6H11ClO2
150.60
MFCD00273371
BNPOTXLWPZOESZ-UHFFFAOYNA-N
r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, r-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4r-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, 4r, r-3-chloro-1,2-propanediol acetonide, pubchem14074, ksc911s2r
2734443
(4R)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
CC1(C)OCC(CCl)O1
Density 1.1030g/mL
Boiling Point 63°C (37.0 mmHg)
Flash Point 51°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.103
Formula Weight 150.6
Percent Purity 98%
Chemical Name or Material (R)-(+)-4-(Chloromethyl)-2, 2-dimethyl-1, 3-dioxolane
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (R)-(+)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'