missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Hydroxypiperidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-4228725
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
198976-43-1
C5H12ClNO
137.61
MFCD00192221
VLECDMDGMKPUSK-NUBCRITNSA-N
r-3-hydroxypiperidine hydrochloride, r-piperidin-3-ol hydrochloride, 3r-piperidin-3-ol hydrochloride, r-+-3-hydroxypiperidine hydrochloride, r-3-hydroxypiperidine hydrocloride, r-3-hydroxypiperidine hcl, r-+-3-piperidinol hydrochloride, r-3-hydroxylpiperidine hydrochloride, 3r-3-piperidinol hydrochloride, r-3-piperidinol hydrochloride
2759333
(3R)-piperidin-3-ol;hydrochloride
C1CC(CNC1)O.Cl
This item is not returnable. View return policy
198976-43-1
C5H12ClNO
137.61
MFCD00192221
VLECDMDGMKPUSK-NUBCRITNSA-N
r-3-hydroxypiperidine hydrochloride, r-piperidin-3-ol hydrochloride, 3r-piperidin-3-ol hydrochloride, r-+-3-hydroxypiperidine hydrochloride, r-3-hydroxypiperidine hydrocloride, r-3-hydroxypiperidine hcl, r-+-3-piperidinol hydrochloride, r-3-hydroxylpiperidine hydrochloride, 3r-3-piperidinol hydrochloride, r-3-piperidinol hydrochloride
2759333
(3R)-piperidin-3-ol;hydrochloride
C1CC(CNC1)O.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 134°C to 138°C
Color White
Elemental Analysis 42.18 to 44.73%, 9.92 to 10.43%
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 1 g
Specific Rotation + 7.1
Formula Weight 137.61
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-3-Hydroxypiperidine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-3-Hydroxypiperidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'