missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Fmoc-amino)-3-phenylpropionic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7034391
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
220498-02-2
C24H21NO4
387.435
MFCD01311783
PTSLRPMRTOVHAB-JOCHJYFZSA-N
r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-3-amino-3-phenylpropionic acid, fmoc-l-beta-phenylalanine, fmoc-beta-phe-oh, r-n-fmoc-3-amino-3-phenylpropanoic acid, r-3-fmoc-amino-3-phenylpropionic acid, fmoc-r-phenylalanine, 3r-3-9h-fluoren-9-ylmethoxycarbonylamino-3-phenylpropanoic acid, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoic acid, fmoc-d-?-phe-oh
2734302
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
This item is not returnable. View return policy
220498-02-2
C24H21NO4
387.435
MFCD01311783
PTSLRPMRTOVHAB-JOCHJYFZSA-N
r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-3-amino-3-phenylpropionic acid, fmoc-l-beta-phenylalanine, fmoc-beta-phe-oh, r-n-fmoc-3-amino-3-phenylpropanoic acid, r-3-fmoc-amino-3-phenylpropionic acid, fmoc-r-phenylalanine, 3r-3-9h-fluoren-9-ylmethoxycarbonylamino-3-phenylpropanoic acid, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoic acid, fmoc-d-?-phe-oh
2734302
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 7982022
Formula Weight 387.43
Percent Purity 95%
Chemical Name or Material (R)-3-(Fmoc-amino)-3-phenylpropionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'