Learn More
(R)-3-Carboxy-4-hydroxyphenylglycine, Tocris Bioscience™
Ionotropic glutamate receptor antagonist
Brand:  Tocris (Bio-Techne) 032850
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13487553
  • £1240.26 / 50mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Moderately potent NMDA and AMPA/kainate receptor antagonist. (S)-3-Carboxy-4-hydroxyphenylglycine (Cat. No. 0329) also available.
Specifications
Chemical Identifiers
13861-03-5
211.173
6604713
C9H9NO5
CHZBCZTXSTWCIG-SSDOTTSWSA-N
Specifications
(R)-3-Carboxy-4-hydroxyphenylglycine
>98%
CHZBCZTXSTWCIG-SSDOTTSWSA-N
5-[(R)-amino(carboxy)methyl]-2-hydroxybenzoic acid
6604713
50mg
13861-03-5
C9H9NO5
C1=CC(=C(C=C1C(C(=O)O)N)C(=O)O)O
211.173
211.17
Videos